Please login to your online account to display your discounted pricing

6-Nitropiperonal, 98+%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC160070050

 View more versions of this product

Catalog No. AC160070050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 195.13
Physical Form Fine Crystalline Powder
Quantity 5g
Color Yellow
RTECS Number DF4912750
EINECS Number 211-926-1
Melting Point 92° to 98°C
CAS Max % 100.0
CAS Min % 98.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C8H5NO5
CAS 712-97-0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Name Note 98+%
Assay Percent Range 98+%
Chemical Name or Material 6-Nitropiperonal
MDL Number MFCD00005829

Specifications

Chemical Name or Material 6-Nitropiperonal
Name Note 98+%
CAS 712-97-0
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C8H5NO5
Formula Weight 195.13
MDL Number MFCD00005829
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents