Please login to your online account to display your discounted pricing

6-Nitropiperonal, 98+%, ACROS Organics™


Quantity 100g

Manufacturer: fisher scientific acros  160071000

 View more versions of this product

Catalog No. AC160071000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Fine Crystalline Powder
Formula Weight 195.13
Quantity 100g
Color Yellow
RTECS Number DF4912750
EINECS Number 211-926-1
Melting Point 92° to 98°C
CAS Max % 100.0
CAS Min % 98.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C8H5NO5
Chemical Name or Material 6-Nitropiperonal
CAS 712-97-0
Assay Percent Range 98+%
MDL Number MFCD00005829
Name Note 98+%

Specifications

Chemical Name or Material 6-Nitropiperonal
Name Note 98+%
CAS 712-97-0
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C8H5NO5
Formula Weight 195.13
MDL Number MFCD00005829
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents