Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Nitropiperonal, 98+%

$125.07 - $262.18

Chemical Identifiers

Chemical Name or Material 6-Nitropiperonal
CAS 712-97-0
Molecular Formula C8H5NO5
Formula Weight 195.13
MDL Number MFCD00005829
Beilstein 384010
Synonym 4,5-Methylenedioxy-2-nitrobenzaldehyde; 6-Nitro-1,3-benzodioxole-5-carboxaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1403914

 
alfa aesar™
A1403914
25g Each for $125.07

AAA1403918

 
alfa aesar™
A1403918
50g Each for $262.18
Description & Specifications

Specifications

Formula Weight 195.13
CAS 712-97-0
Molecular Formula C8H5NO5
Beilstein 384010
Chemical Name or Material 6-Nitropiperonal
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 4,5-Methylenedioxy-2-nitrobenzaldehyde; 6-Nitro-1,3-benzodioxole-5-carboxaldehyde
MDL Number MFCD00005829
Assay ≥98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 93° to 97°C
Quantity 25g
RTECS Number DF4912750
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.