Please login to your online account to display your discounted pricing

6-Nitrophthalide, 97%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  331950100

 View more versions of this product

Catalog No. AC331950100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 179.13
Quantity 10g
Color Pale yellow
Beilstein 17, 313
Melting Point 140° to 145°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 6-Nitrophthalide
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C8H5NO4
MDL Number MFCD00033529
CAS 610-93-5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 6-Nitrophthalide
Name Note 97%
CAS 610-93-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H5NO4
Formula Weight 179.13
MDL Number MFCD00033529