Please login to your online account to display your discounted pricing

6-Nitrophthalide, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 6-Nitrophthalide
Name Note 97%
CAS 610-93-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H5NO4
Formula Weight 179.13
MDL Number MFCD00033529
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC331950100

  SDS
fisher scientific acros
331950100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC331950010

  SDS
thermo fisher scientific
331950010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 179.13
Quantity 10g
Color Pale yellow
Beilstein 17, 313
Melting Point 140° to 145°C
CAS Max % 100.0
CAS Min % 96.0
Infrared Spectrum Authentic
Chemical Name or Material 6-Nitrophthalide
Packaging Glass bottle
Name Note 97%
MDL Number MFCD00033529
Molecular Formula C8H5NO4
Assay Percent Range 97%
CAS 610-93-5