missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Nitroindoline 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 128710010

 View more versions of this product

Catalog No. AC128710010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

6-Nitroindoline
66° to 70°C
98%
97.5
Authentic
Crystalline Powder or Crystals
98%
164.16
Orange or red
1g
19727-83-4
100.0
Glass bottle
≥97.5%
C8H8N2O2
MFCD00005710

Chemical Identifiers

19727-83-4
100.0
C8H8N2O2
MFCD00005710
≥97.5%
97.5
98%
164.16
6-Nitroindoline, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 243-257-6

RTECSNumber : NM1950000