Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Nitroindoline, 98+%

$100.96 - $400.80

Chemical Identifiers

Chemical Name or Material 6-Nitroindoline
CAS 19727-83-4
Molecular Formula C8H8N2O2
Formula Weight 164.16
MDL Number MFCD00005710
Beilstein 156434
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2196106

  SDS
alfa aesar™
B2196106
5g Each for $100.96

AAB2196114

  SDS
alfa aesar™
B2196114
25g Each for $400.80
Description & Specifications

Specifications

Formula Weight 164.16
Quantity 5g
Assay ≥98%
Molecular Formula C8H8N2O2
MDL Number MFCD00005710
Health Hazard 1 Danger
Chemical Name or Material 6-Nitroindoline
CAS 19727-83-4
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number NM1950000
Melting Point 66° to 70°C
Health Hazard 2 GHS H Statement
H301-H311-H341-H332
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing genetic defects.
Harmful if inhaled.
Beilstein 156434
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H341-H332
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing genetic defects.
Harmful if inhaled.