Please login to your online account to display your discounted pricing

6-Nitroindole, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC348100050

 View more versions of this product

Catalog No. AC348100050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95% (GC)
Formula Weight 162.15
Physical Form Powder
Quantity 5g
Color Yellow to brown-yellow
Melting Point 137° to 143°C
Assay Percent Range 96%
CAS 4769-96-4
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 6-Nitroindole
Molecular Formula C8H6N2O2

Specifications

Chemical Name or Material 6-Nitroindole
Assay ≥95% (GC)
Assay Percent Range 96%
Molecular Formula C8H6N2O2
Formula Weight 162.15