Please login to your online account to display your discounted pricing

6-Nitroindole, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 6-Nitroindole
Assay ≥95% (GC)
Assay Percent Range 96%
Molecular Formula C8H6N2O2
Formula Weight 162.15
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC348100050

  SDS
thermo fisher scientific
AC348100050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 6-Nitroindole
Color Yellow to brown-yellow
Melting Point 137° to 143°C
Quantity 5g
CAS 4769-96-4
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Powder
Assay ≥95% (GC)
Assay Percent Range 96%
Molecular Formula C8H6N2O2
Formula Weight 162.15