missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Nitro-2H-1,4-benzothiazin-3(4H)-one, 97%

$52.86 - $167.51

Chemical Identifiers

CAS 21762-74-3
Molecular Formula C8H6N2O3S
Molecular Weight (g/mol) 210.207
MDL Number MFCD00456556
InChI Key BALSKAVZBPPKCS-UHFFFAOYSA-N
Synonym 2h-1,4-benzothiazin-3 4h-one, 6-nitro
PubChem CID 4182356
IUPAC Name 6-nitro-4H-1,4-benzothiazin-3-one
SMILES C1C(=O)NC2=C(S1)C=CC(=C2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61031MD View Documents Alfa Aesar™
H61031MD
250mg Each for $52.86
 
AAH6103103 View Documents Alfa Aesar™
H6103103
1g Each for $167.51
 
Specifications

Chemical Identifiers

21762-74-3
210.207
BALSKAVZBPPKCS-UHFFFAOYSA-N
4182356
C1C(=O)NC2=C(S1)C=CC(=C2)[N+](=O)[O-]
C8H6N2O3S
MFCD00456556
2h-1,4-benzothiazin-3 4h-one, 6-nitro
6-nitro-4H-1,4-benzothiazin-3-one

Specifications

250mg
C8H6N2O3S
2h-1,4-benzothiazin-3 4h-one, 6-nitro
C1C(=O)NC2=C(S1)C=CC(=C2)[N+](=O)[O-]
210.207
210.21
6-Nitro-2H-1,4-benzothiazin-3(4H)-one
21762-74-3
MFCD00456556
BALSKAVZBPPKCS-UHFFFAOYSA-N
6-nitro-4H-1,4-benzothiazin-3-one
4182356
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures