missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Nitro-2H-1,4-benzothiazin-3(4H)-one, 97%

Manufacturer:  Alfa Aesar™ H6103103

 View more versions of this product

Catalog No. AAH6103103



Specifications

Specifications

6-Nitro-2H-1,4-benzothiazin-3(4H)-one
21762-74-3
MFCD00456556
BALSKAVZBPPKCS-UHFFFAOYSA-N
6-nitro-4H-1,4-benzothiazin-3-one
4182356
97%
1g
C8H6N2O3S
2h-1,4-benzothiazin-3 4h-one, 6-nitro
C1C(=O)NC2=C(S1)C=CC(=C2)[N+](=O)[O-]
210.207
210.21

Chemical Identifiers

21762-74-3
210.207
BALSKAVZBPPKCS-UHFFFAOYSA-N
4182356
C1C(=O)NC2=C(S1)C=CC(=C2)[N+](=O)[O-]
C8H6N2O3S
MFCD00456556
2h-1,4-benzothiazin-3 4h-one, 6-nitro
6-nitro-4H-1,4-benzothiazin-3-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents