Learn More
6-Nitro-1H-indazole, 97%, Thermo Scientific Chemicals
$70.23 - $601.67
Chemical Identifiers
CAS | 7597-18-4 |
---|---|
Molecular Formula | C7H5N3O2 |
Molecular Weight (g/mol) | 163.136 |
MDL Number | MFCD00005695 |
InChI Key | ORZRMRUXSPNQQL-UHFFFAOYSA-N |
Synonym | 6-nitroindazole, 6-nitro-2h-indazole, 6-nitroisoindazole, 1h-indazole, 6-nitro, 2h-indazole, 6-nitro, indazole,6-nitro, ccris 3263, chembl54277, 6ni, 6-nitro-indazol |
PubChem CID | 24239 |
IUPAC Name | 6-nitro-1H-indazole |
SMILES | C1=CC2=C(C=C1[N+](=O)[O-])NN=C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1621314
|
Thermo Scientific Chemicals
A1621314 |
25 g |
N/A
|
|
|||||
AAA1621322
|
Thermo Scientific Chemicals
A1621322 |
100 g |
N/A
|
|
|||||
AAA1621336
|
Thermo Scientific Chemicals
A1621336 |
500 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7597-18-4 | |
163.136 | |
ORZRMRUXSPNQQL-UHFFFAOYSA-N | |
24239 | |
C1=CC2=C(C=C1[N+](=O)[O-])NN=C2 |
C7H5N3O2 | |
MFCD00005695 | |
6-nitroindazole, 6-nitro-2h-indazole, 6-nitroisoindazole, 1h-indazole, 6-nitro, 2h-indazole, 6-nitro, indazole,6-nitro, ccris 3263, chembl54277, 6ni, 6-nitro-indazol | |
6-nitro-1H-indazole |
Specifications
7597-18-4 | |
Odorless | |
MFCD00005695 | |
7812 | |
ORZRMRUXSPNQQL-UHFFFAOYSA-N | |
6-nitro-1H-indazole | |
24239 | |
97% |
179°C to 182°C | |
C7H5N3O2 | |
25 g | |
6-nitroindazole, 6-nitro-2h-indazole, 6-nitroisoindazole, 1h-indazole, 6-nitro, 2h-indazole, 6-nitro, indazole,6-nitro, ccris 3263, chembl54277, 6ni, 6-nitro-indazol | |
C1=CC2=C(C=C1[N+](=O)[O-])NN=C2 | |
163.136 | |
163.14 | |
6-Nitro-1H-indazole |
Safety and Handling
GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H340-H350
EINECSNumber : 231-500-9
RTECSNumber : NK7962100
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only