Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(N-Boc-methylamino)-5-methylpyridine-3-boronic acid pinacol ester, 95%

$224.51 - $515.31

Chemical Identifiers

Chemical Name or Material 6-(N-Boc-methylamino)-5-methylpyridine-3-boronic acid pinacol ester
CAS 1032758-82-9
Molecular Formula C18H29BN2O4
Formula Weight 348.25
MDL Number MFCD09037485
Synonym tert-Butyl N-methyl-N-[3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridyl]carbamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2750003

  SDS
alfa aesar™
H2750003
1g Each for $515.31

AAH27500MD

  SDS
alfa aesar™
H27500MD
250mg Each for $224.51
Description & Specifications

Specifications

Formula Weight 348.25
Chemical Name or Material 6-(N-Boc-methylamino)-5-methylpyridine-3-boronic acid pinacol ester
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD09037485
Melting Point 152° to 155°C
Synonym tert-Butyl N-methyl-N-[3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridyl]carbamate
CAS 1032758-82-9
Molecular Formula C18H29BN2O4
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Assay 95%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.