Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(N,2,2,2-Tetramethylacetamido)-5-methylpyridine-3-boronic acid pinacol ester, 95%

$224.45 - $224.45

Chemical Identifiers

Chemical Name or Material 6-(N,2,2,2-Tetramethylacetamido)-5-methylpyridine-3-boronic acid pinacol ester
CAS 1073354-77-4
Molecular Formula C18H29BN2O3
Formula Weight 332.25g/mol
MDL Number MFCD09037482
Synonym 6-(N-Methylpivalamido)-5-methylpyridine-3-boronic acid pinacol ester; N-[3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridyl]-N-methyl-2,2-dimethylpropionamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27211MD

 
alfa aesar™
H27211-MD
250mg Each for $224.45
Description & Specifications

Specifications

Melting Point 134° to 136°C
Quantity 250mg
Chemical Name or Material 6-(N,2,2,2-Tetramethylacetamido)-5-methylpyridine-3-boronic acid pinacol ester
CAS 1073354-77-4
Assay 95%
Molecular Formula C18H29BN2O3
Formula Weight 332.25g/mol
MDL Number MFCD09037482
Synonym 6-(N-Methylpivalamido)-5-methylpyridine-3-boronic acid pinacol ester; N-[3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridyl]-N-methyl-2,2-dimethylpropionamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.