Please login to your online account to display your discounted pricing

6-Morpholinonicotinic acid, 95+%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC17501DE

 View more versions of this product

Catalog No. CC17501DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 208.22
Quantity 5g
Assay 95%
Chemical Name or Material 6-Morpholinonicotinic acid
Assay Percent Range 95%
Molecular Formula C10H12N2O3
CAS 120800-52-4
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 6-Morpholinonicotinic acid
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H12N2O3
Formula Weight 208.22