missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(6-Morpholino-3-pyridinyl)methanol, 95%, Maybridge

Manufacturer:  Thermo Scientific CC17509DA

 View more versions of this product

Catalog No. CC17509DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(6-Morpholino-3-pyridinyl)methanol
1g
94.5
95%
MFCD03086115
NXKXCMMRCZMJRJ-UHFFFAOYSA-N
(6-morpholin-4-ylpyridin-3-yl)methanol
2776465
95%
Amber Glass Bottle
388088-73-1
100.0
C10H14N2O2
6-morpholino-3-pyridinyl methanol, 6-morpholin-4-yl pyridin-3-yl methanol, 6-morpholinopyridin-3-yl methanol, 5-hydroxymethyl-2-morpholin-4-ylpyridine, 3-pyridinemethanol,6-4-morpholinyl, 6-morpholin-4-ylpyridin-3-yl methanol, 6-morpholin-4-yl-3-pyridyl methan-1-ol, 6-morpholin-4-yl pyridine-3-yl methanol
C1COCCN1C2=NC=C(C=C2)CO
194.234
194.23

Chemical Identifiers

388088-73-1
194.234
NXKXCMMRCZMJRJ-UHFFFAOYSA-N
2776465
C1COCCN1C2=NC=C(C=C2)CO
C10H14N2O2
MFCD03086115
6-morpholino-3-pyridinyl methanol, 6-morpholin-4-yl pyridin-3-yl methanol, 6-morpholinopyridin-3-yl methanol, 5-hydroxymethyl-2-morpholin-4-ylpyridine, 3-pyridinemethanol,6-4-morpholinyl, 6-morpholin-4-ylpyridin-3-yl methanol, 6-morpholin-4-yl-3-pyridyl methan-1-ol, 6-morpholin-4-yl pyridine-3-yl methanol
(6-morpholin-4-ylpyridin-3-yl)methanol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents