missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC77751DE
Specifications
6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate | |
Amber Glass Bottle | |
5g | |
VARLCSQEQQKXKH-UHFFFAOYSA-N | |
6-morpholin-4-ylpyridazine-3-carboxylic acid;hydrate;hydrochloride | |
53444630 | |
97% |
1192758-40-9 | |
C9H14ClN3O4 | |
6-morpholinopyridazine-3-carboxylic acid hydrochloride hydrate, 6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate, 6-morpholin-4-yl pyridazine-3-carboxylic acid hydrate hydrochloride | |
C1COCCN1C2=NN=C(C=C2)C(=O)O.O.Cl | |
263.678 | |
245.67 |
Chemical Identifiers
1192758-40-9 | |
263.678 | |
6-morpholinopyridazine-3-carboxylic acid hydrochloride hydrate, 6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate, 6-morpholin-4-yl pyridazine-3-carboxylic acid hydrate hydrochloride | |
6-morpholin-4-ylpyridazine-3-carboxylic acid;hydrate;hydrochloride |
C9H14ClN3O4 | |
VARLCSQEQQKXKH-UHFFFAOYSA-N | |
53444630 | |
C1COCCN1C2=NN=C(C=C2)C(=O)O.O.Cl |