Please login to your online account to display your discounted pricing

6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC77751DA

 View more versions of this product

Catalog No. CC77751DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 245.67
Quantity 1g
Assay 97%
Molecular Formula C9H11N3O3·ClH
CAS 1192758-40-9
Packaging Amber Glass Bottle
Chemical Name or Material 6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate

Specifications

Chemical Name or Material 6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate
Assay 97%
Molecular Formula C9H11N3O3·ClH
Formula Weight 245.67