missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(Methylthio)purine, 97%

$285.30 - $285.30

Chemical Identifiers

CAS 50-66-8
Molecular Formula C6H6N4S
Molecular Weight (g/mol) 166.202
MDL Number MFCD00005576
InChI Key UIJIQXGRFSPYQW-UHFFFAOYSA-N
Synonym 6-methylthio purine, 6-methylmercaptopurine, 6-methylthiopurine, 1h-purine, 6-methylthio, 6-methyl mp, purine, 6-methylthio, s-methyl-6-mercaptopurine, 6-methylsulfanyl-9h-purine, unii-6v404dv25o, thiopurine s-methylether
PubChem CID 5778
ChEBI CHEBI:28279
IUPAC Name 6-methylsulfanyl-7H-purine
SMILES CSC1=NC=NC2=C1NC=N2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0183606 View Documents Alfa Aesar™
L0183606
5g Each for $285.30
 
Description

Description

Applications
6-(Methylthio)purine can react with 1-bromo-2-chloro-ethane to produce 6-methylsulfanyl-9-vinyl-9H-purine. This reaction will need reagent NaOH, catalyst (C8H17)4NBr, and the menstruum benzene and H2O.

Solubility
Soluble in water (769 mg/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

50-66-8
166.202
UIJIQXGRFSPYQW-UHFFFAOYSA-N
5778
6-methylsulfanyl-7H-purine
C6H6N4S
MFCD00005576
6-methylthio purine, 6-methylmercaptopurine, 6-methylthiopurine, 1h-purine, 6-methylthio, 6-methyl mp, purine, 6-methylthio, s-methyl-6-mercaptopurine, 6-methylsulfanyl-9h-purine, unii-6v404dv25o, thiopurine s-methylether
CHEBI:28279
CSC1=NC=NC2=C1NC=N2

Specifications

220°C to 223°C
50-66-8
MFCD00005576
6-methylthio purine, 6-methylmercaptopurine, 6-methylthiopurine, 1h-purine, 6-methylthio, 6-methyl mp, purine, 6-methylthio, s-methyl-6-mercaptopurine, 6-methylsulfanyl-9h-purine, unii-6v404dv25o, thiopurine s-methylether
CSC1=NC=NC2=C1NC=N2
166.202
CHEBI:28279
97%
5g
C6H6N4S
7695
UIJIQXGRFSPYQW-UHFFFAOYSA-N
6-methylsulfanyl-7H-purine
5778
166.2
6-(Methylthio)purine
Safety and Handling

Safety and Handling

RTECSNumber : UO8976000