Please login to your online account to display your discounted pricing

6-(Methylsulfanyl)-7H-purin-2-ylamine, 97%, ACROS Organics™


Quantity 50mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC104020500

 View more versions of this product

Catalog No. AC104020500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 181.22
Quantity 50mg
Color Off-white
Color Index No. 2-Amino-6-methylmercaptopurine
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 2-Amino-6-methylmercaptopurine
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 95%
Molecular Formula C6H7N5S
MDL Number MFCD00044701
CAS 1198-47-6
CAS 1198-47-6

Specifications

Chemical Name or Material 2-Amino-6-methylmercaptopurine
CAS 1198-47-6
CAS Min % 94.0
CAS Max % 100.0
CAS 1198-47-6
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H7N5S
Formula Weight 181.22
MDL Number MFCD00044701