Please login to your online account to display your discounted pricing

6-(Methylsulfanyl)-7H-purin-2-ylamine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Amino-6-methylmercaptopurine
CAS 1198-47-6
CAS Min % 94.0
CAS Max % 100.0
CAS 1198-47-6
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C6H7N5S
Formula Weight 181.22g/mol
MDL Number MFCD00044701
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC104020500

  SDS
thermo fisher scientific
104020500
Glass bottle 50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color Index No. 2-Amino-6-methylmercaptopurine
Color Off-white
Quantity 50mg
Chemical Name or Material 2-Amino-6-methylmercaptopurine
CAS 1198-47-6
CAS Min % 94.0
CAS Max % 100.0
CAS 1198-47-6
Physical Form Off-white crystalline powder
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C6H7N5S
Formula Weight 181.22g/mol
MDL Number MFCD00044701