Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methylquinoxaline, 96%

$77.91 - $77.91

Chemical Identifiers

Chemical Name or Material 6-Methylquinoxaline
CAS 6344-72-5
Molecular Formula C9H8N2
Formula Weight 144.18
MDL Number MFCD00041001
Beilstein 113371
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1109003

  SDS
alfa aesar™
L1109003
1g Each for $77.91
Description & Specifications

Specifications

Formula Weight 144.18
Molecular Formula C9H8N2
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Chemical Name or Material 6-Methylquinoxaline
Assay 96%
MDL Number MFCD00041001
CAS 6344-72-5
Beilstein 113371
Boiling Point 244° to 245°C
Health Hazard 1 Warning
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.