missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methylquinoxaline, 96%

Manufacturer:  Alfa Aesar™ L1109006

 View more versions of this product

Catalog No. AAL1109006



Specifications

Specifications

6-Methylquinoxaline
5g
C9H8N2
113371
OSRARURJYPOUOV-UHFFFAOYSA-N
6-methylquinoxaline
242567
96%
244°C to 245°C
6344-72-5
MFCD00041001
quinoxaline, 6-methyl, 6-methyl quinoxaline, 6-methyl-quinoxaline, acmc-1b9iq, ksc356q6h
CC1=CC2=NC=CN=C2C=C1
144.177
144.18

Chemical Identifiers

6344-72-5
144.177
OSRARURJYPOUOV-UHFFFAOYSA-N
242567
CC1=CC2=NC=CN=C2C=C1
C9H8N2
MFCD00041001
quinoxaline, 6-methyl, 6-methyl quinoxaline, 6-methyl-quinoxaline, acmc-1b9iq, ksc356q6h
6-methylquinoxaline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents