missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methylquinoline 98.0+%, TCI America™

Manufacturer:  TCI America M041625G

 View more versions of this product

Catalog No. M041625G



Specifications

Specifications

6-Methylquinoline
Yellow
91-62-3
MFCD00006804
LUYISICIYVKBTA-UHFFFAOYSA-N
6-methylquinoline
7059
Clear Liquid at 20°C
259°C
25g
C10H9N
quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887
CC1=CC2=C(C=C1)N=CC=C2
143.189
143.19
≥98.0% (GC,T)

Chemical Identifiers

91-62-3
143.189
LUYISICIYVKBTA-UHFFFAOYSA-N
7059
CC1=CC2=C(C=C1)N=CC=C2
C10H9N
MFCD00006804
quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887
6-methylquinoline
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0796

RTECSNumber : VC0550000

TSCA : Yes

Documents