Learn More
6-Methylquinoline, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 295140250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
6-Methylquinoline | |
1.0600g/mL | |
>113°C | |
97.5% min. (GC) | |
C10H9N | |
MFCD00006804 | |
1.06 | |
Solubility in water: .. Other solubilities: soluble in ethanol | |
CC1=CC=C2N=CC=CC2=C1 | |
143.19 | |
143.19 |
91-62-3 | |
256.0°C to 260.0°C | |
Authentic | |
Glass bottle | |
1.6125 to 1.6145 | |
25g | |
quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887 | |
LUYISICIYVKBTA-UHFFFAOYSA-N | |
6-methylquinoline | |
7059 | |
98% |
Chemical Identifiers
91-62-3 | |
143.19 | |
LUYISICIYVKBTA-UHFFFAOYSA-N | |
7059 | |
CC1=CC=C2N=CC=CC2=C1 |
C10H9N | |
MFCD00006804 | |
quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887 | |
6-methylquinoline |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before re
GHS Signal Word: Warning
EINECSNumber : 202-084-6
RUO â Research Use Only