missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methylquinazoline-2,4(1H,3H)-dione, 97%

Manufacturer:  Alfa Aesar™ H6325906

 View more versions of this product

Catalog No. AAH6325906



Specifications

Specifications

6-Methylquinazoline-2,4(1H,3H)-dione
5g
C9H8N2O2
6-methylquinazoline-2,4 1h,3h-dione, 6-methylquinazoline-2,4-diol, 2,4 1h,3h-quinazolinedione, 6-methyl, 6-methyl-1h-quinazoline-2,4-quinone, 6-methyl-2,4 1h,3h-quinazolinedione, 6-methylquinazoline-2,4-1h,3h-dione, 2,4-dihydroxy-6-methylquinazoline, quinazoline-2,4-dione,6-me, 6-methyl-2,4-1h,3h-quinazolinedione, 2,4 1h,3h-quinazolinedione,6-methyl
CC1=CC2=C(C=C1)NC(=O)NC2=O
176.175
176.17
97%
White
62484-16-6
MFCD00831482
OXVPNGBFUAIXTE-UHFFFAOYSA-N
6-methyl-1H-quinazoline-2,4-dione
334024
Solid

Chemical Identifiers

62484-16-6
176.175
OXVPNGBFUAIXTE-UHFFFAOYSA-N
334024
CC1=CC2=C(C=C1)NC(=O)NC2=O
C9H8N2O2
MFCD00831482
6-methylquinazoline-2,4 1h,3h-dione, 6-methylquinazoline-2,4-diol, 2,4 1h,3h-quinazolinedione, 6-methyl, 6-methyl-1h-quinazoline-2,4-quinone, 6-methyl-2,4 1h,3h-quinazolinedione, 6-methylquinazoline-2,4-1h,3h-dione, 2,4-dihydroxy-6-methylquinazoline, quinazoline-2,4-dione,6-me, 6-methyl-2,4-1h,3h-quinazolinedione, 2,4 1h,3h-quinazolinedione,6-methyl
6-methyl-1H-quinazoline-2,4-dione
Documents