Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methylpyridine-3-carboxaldehyde, 96%

Chemical Identifiers

Chemical Name or Material 6-Methylpyridine-3-carboxaldehyde
CAS 53014-84-9
Formula Weight 121.14
MDL Number MFCD08272279
Beilstein 107496
Synonym 6-Methylnicotinaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6692606

 
alfa aesar™
H6692606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6692603

 
alfa aesar™
H6692603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66926MD

 
alfa aesar™
H66926MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 121.14
Chemical Name or Material 6-Methylpyridine-3-carboxaldehyde
CAS 53014-84-9
Physical Form Liquid
Assay 96%
MDL Number MFCD08272279
Beilstein 107496
Synonym 6-Methylnicotinaldehyde
Refractive Index 1.5460
Boiling Point 95°C (20mmHg)
Density 1.084g/mL
Sensitivity Moisture Sensitive
Quantity 5g