missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Methylnicotinamide 98.0+%, TCI America™
Supplier: TCI America M247725G
Specifications
6-Methylnicotinamide | |
199°C | |
C7H8N2O | |
25 g | |
IJXDURUAYOKSIS-UHFFFAOYSA-N | |
6-methylpyridine-3-carboxamide | |
96351 | |
≥98.0% (HPLC,T) |
6960-22-1 | |
White-Yellow | |
MFCD00006342 | |
6-methylnicotinamide, 6-methyl nicotinamide, 3-pyridinecarboxamide, 6-methyl, 6-methyl-3-pyridinecarboxamide, 8ej, nicotinamide, 6-methyl, 3-carbamoyl-6-methylpyridine, ksc630k7f, 2-methylpyridine-5-carboxamide, 3-pyridinecarboxamide,6-methyl | |
CC1=NC=C(C=C1)C(=O)N | |
136.154 | |
136.15 | |
Crystalline Powder |
Chemical Identifiers
6960-22-1 | |
136.154 | |
IJXDURUAYOKSIS-UHFFFAOYSA-N | |
96351 | |
CC1=NC=C(C=C1)C(=O)N |
C7H8N2O | |
MFCD00006342 | |
6-methylnicotinamide, 6-methyl nicotinamide, 3-pyridinecarboxamide, 6-methyl, 6-methyl-3-pyridinecarboxamide, 8ej, nicotinamide, 6-methyl, 3-carbamoyl-6-methylpyridine, ksc630k7f, 2-methylpyridine-5-carboxamide, 3-pyridinecarboxamide,6-methyl | |
6-methylpyridine-3-carboxamide |
Safety and Handling
TSCA : No