missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methylindole-3-carboxaldehyde, 98%

$55.82 - $165.01

Chemical Identifiers

CAS 4771-49-7
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00049347
InChI Key LZERQSJGPXFAKB-UHFFFAOYSA-N
Synonym 6-methylindole-3-carboxaldehyde, 6-methylindole-3-carboxyaldehyde, 1h-indole-3-carboxaldehyde, 6-methyl, 6-methylindole-3-carbaldehyde, 6-methyl-1h-indole-3-carboxaldehyde, 1h-indole-3-carboxaldehyde,6-methyl, pubchem7241, acmc-209ka7, ksc497o2l, 6-methylindole-3-caboxaldehyde
PubChem CID 4777902
IUPAC Name 6-methyl-1H-indole-3-carbaldehyde
SMILES CC1=CC2=C(C=C1)C(=CN2)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1750803 View Documents Alfa Aesar™
L1750803
1g Each for $165.01
 
AAL17508MD View Documents Alfa Aesar™
L17508MD
250mg Each for $55.82
 
Specifications

Chemical Identifiers

4771-49-7
159.188
LZERQSJGPXFAKB-UHFFFAOYSA-N
4777902
CC1=CC2=C(C=C1)C(=CN2)C=O
C10H9NO
MFCD00049347
6-methylindole-3-carboxaldehyde, 6-methylindole-3-carboxyaldehyde, 1h-indole-3-carboxaldehyde, 6-methyl, 6-methylindole-3-carbaldehyde, 6-methyl-1h-indole-3-carboxaldehyde, 1h-indole-3-carboxaldehyde,6-methyl, pubchem7241, acmc-209ka7, ksc497o2l, 6-methylindole-3-caboxaldehyde
6-methyl-1H-indole-3-carbaldehyde

Specifications

185°C to 189°C
4771-49-7
MFCD00049347
6-methylindole-3-carboxaldehyde, 6-methylindole-3-carboxyaldehyde, 1h-indole-3-carboxaldehyde, 6-methyl, 6-methylindole-3-carbaldehyde, 6-methyl-1h-indole-3-carboxaldehyde, 1h-indole-3-carboxaldehyde,6-methyl, pubchem7241, acmc-209ka7, ksc497o2l, 6-methylindole-3-caboxaldehyde
CC1=CC2=C(C=C1)C(=CN2)C=O
159.188
159.19
6-Methylindole-3-carboxaldehyde
1g
C10H9NO
386914
LZERQSJGPXFAKB-UHFFFAOYSA-N
6-methyl-1H-indole-3-carbaldehyde
4777902
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning