missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Methylflavone 98.0+%, TCI America™
$132.71 - $462.86
Chemical Identifiers
CAS | 29976-75-8 |
---|---|
Molecular Formula | C16H12O2 |
Molecular Weight (g/mol) | 236.27 |
MDL Number | MFCD00017461 |
InChI Key | NOQJBXPAMJLUSS-UHFFFAOYSA-N |
PubChem CID | 689013 |
IUPAC Name | 6-methyl-2-phenylchromen-4-one |
SMILES | CC1=CC2=C(C=C1)OC(=CC2=O)C3=CC=CC=C3 |
Chemical Identifiers
29976-75-8 | |
236.27 | |
NOQJBXPAMJLUSS-UHFFFAOYSA-N | |
6-methyl-2-phenylchromen-4-one |
C16H12O2 | |
MFCD00017461 | |
689013 | |
CC1=CC2=C(C=C1)OC(=CC2=O)C3=CC=CC=C3 |
Specifications
29976-75-8 | |
White-Yellow | |
C16H12O2 | |
1 g | |
CC1=CC2=C(C=C1)OC(=CC2=O)C3=CC=CC=C3 | |
236.27 | |
236.27 | |
Crystalline Powder |
121°C | |
235°C | |
MFCD00017461 | |
NOQJBXPAMJLUSS-UHFFFAOYSA-N | |
6-methyl-2-phenylchromen-4-one | |
689013 | |
≥98.0% (GC) | |
6-Methylflavone |
Safety and Handling
TSCA : No