Please login to your online account to display your discounted pricing

6-Methylchromone hydrate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 6-Methylchromone hydrate
CAS 207511-19-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H8O2·xH2O
Formula Weight 160.17
MDL Number MFCD00209598
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC373020050

  SDS
thermo fisher scientific
AC373020050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 160.17
Quantity 5g
Color White
Melting Point 70° to 75°C
CAS Max % 100.0
Chemical Name or Material 6-Methylchromone hydrate
Assay Percent Range 99%
MDL Number MFCD00209598
CAS 207511-19-1
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C10H8O2·xH2O