missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Methylchromone-3-carbonitrile 98.0+%, TCI America™
$26.54 - $63.05
Chemical Identifiers
CAS | 50743-18-5 |
---|---|
Molecular Formula | C11H7NO2 |
Molecular Weight (g/mol) | 185.18 |
MDL Number | MFCD00191962 |
InChI Key | FNRLRLMPBIRUSQ-UHFFFAOYSA-N |
Synonym | 3-cyano-6-methylchromone, 6-methylchromone-3-carbonitrile, 6-methyl-4-oxo-4h-1-benzopyran-3-carbonitrile, 4h-1-benzopyran-3-carbonitrile, 6-methyl-4-oxo, 6-methyl-4-oxo-4h-chromene-3-carbonitrile, acmc-1at8c, 6-methylchromanone-3-carbonitrile, 6-methyl-4-oxo-1-benzopyran-3-carbonitrile, 4-oxo-6-methyl-4h-1-benzopyran-3-carbonitrile |
PubChem CID | 688705 |
IUPAC Name | 6-methyl-4-oxo-4H-chromene-3-carbonitrile |
SMILES | CC1=CC=C2OC=C(C#N)C(=O)C2=C1 |
Chemical Identifiers
50743-18-5 | |
185.18 | |
FNRLRLMPBIRUSQ-UHFFFAOYSA-N | |
688705 | |
CC1=CC=C2OC=C(C#N)C(=O)C2=C1 |
C11H7NO2 | |
MFCD00191962 | |
3-cyano-6-methylchromone, 6-methylchromone-3-carbonitrile, 6-methyl-4-oxo-4h-1-benzopyran-3-carbonitrile, 4h-1-benzopyran-3-carbonitrile, 6-methyl-4-oxo, 6-methyl-4-oxo-4h-chromene-3-carbonitrile, acmc-1at8c, 6-methylchromanone-3-carbonitrile, 6-methyl-4-oxo-1-benzopyran-3-carbonitrile, 4-oxo-6-methyl-4h-1-benzopyran-3-carbonitrile | |
6-methyl-4-oxo-4H-chromene-3-carbonitrile |
Specifications
50743-18-5 | |
Yellow | |
MFCD00191962 | |
3439 | |
FNRLRLMPBIRUSQ-UHFFFAOYSA-N | |
6-methyl-4-oxo-4H-chromene-3-carbonitrile | |
688705 | |
≥98.0% (HPLC,N) | |
6-Methylchromone-3-carbonitrile |
152°C | |
C11H7NO2 | |
1 g | |
3-cyano-6-methylchromone, 6-methylchromone-3-carbonitrile, 6-methyl-4-oxo-4h-1-benzopyran-3-carbonitrile, 4h-1-benzopyran-3-carbonitrile, 6-methyl-4-oxo, 6-methyl-4-oxo-4h-chromene-3-carbonitrile, acmc-1at8c, 6-methylchromanone-3-carbonitrile, 6-methyl-4-oxo-1-benzopyran-3-carbonitrile, 4-oxo-6-methyl-4h-1-benzopyran-3-carbonitrile | |
CC1=CC=C2OC=C(C#N)C(=O)C2=C1 | |
185.18 | |
185.18 | |
Crystalline Powder |
Safety and Handling
TSCA : No