Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methylchromone-3-carbonitrile, 99%

Chemical Identifiers

Chemical Name or Material 6-Methylchromone-3-carbonitrile
CAS 50743-18-5
Molecular Formula C11H7NO2
Formula Weight 185.19
MDL Number MFCD00191962
UN Number UN3439
Synonym 3-Cyano-6-methylchromone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2782503

  SDS
alfa aesar™
H2782503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2782506

  SDS
alfa aesar™
H2782506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 185.19
Chemical Name or Material 6-Methylchromone-3-carbonitrile
Assay 99%
Molecular Formula C11H7NO2
MDL Number MFCD00191962
CAS 50743-18-5
UN Number UN3439
Synonym 3-Cyano-6-methylchromone
Health Hazard 1 Danger
Melting Point 150° to 152°C
Quantity 1g
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.