missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methylchromone-2-carboxylic Acid, TCI America™

$215.42 - $644.73

Chemical Identifiers

CAS 5006-44-0
Molecular Formula C11H8O4
Molecular Weight (g/mol) 204.181
MDL Number MFCD00239435
InChI Key RKIDLZFIIVDZNX-UHFFFAOYSA-N
Synonym 6-methylchromone-2-carboxylic acid, 6-methyl-4-oxo-4h-chromene-2-carboxylic acid, 4h-1-benzopyran-2-carboxylic acid, 6-methyl-4-oxo, 6-methyl-4-oxo-4h-1-benzopyran-2-carboxylic acid, maybridge3_003393, 6-methylchromone-2-carboxylic acid, 4h-1-benzopyran-2-carboxylicacid, 6-methyl-4-oxo
PubChem CID 688704
IUPAC Name 6-methyl-4-oxochromene-2-carboxylic acid
SMILES CC1=CC2=C(C=C1)OC(=CC2=O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M27255G View Documents TCI America
M27255G
5g Each for $644.73
 
M27251G View Documents TCI America
M27251G
1g Each for $215.42
 
Specifications

Chemical Identifiers

5006-44-0
204.181
RKIDLZFIIVDZNX-UHFFFAOYSA-N
688704
CC1=CC2=C(C=C1)OC(=CC2=O)C(=O)O
C11H8O4
MFCD00239435
6-methylchromone-2-carboxylic acid, 6-methyl-4-oxo-4h-chromene-2-carboxylic acid, 4h-1-benzopyran-2-carboxylic acid, 6-methyl-4-oxo, 6-methyl-4-oxo-4h-1-benzopyran-2-carboxylic acid, maybridge3_003393, 6-methylchromone-2-carboxylic acid, 4h-1-benzopyran-2-carboxylicacid, 6-methyl-4-oxo
6-methyl-4-oxochromene-2-carboxylic acid

Specifications

Undesignated
5g
C11H8O4
6-methylchromone-2-carboxylic acid, 6-methyl-4-oxo-4h-chromene-2-carboxylic acid, 4h-1-benzopyran-2-carboxylic acid, 6-methyl-4-oxo, 6-methyl-4-oxo-4h-1-benzopyran-2-carboxylic acid, maybridge3_003393, 6-methylchromone-2-carboxylic acid, 4h-1-benzopyran-2-carboxylicacid, 6-methyl-4-oxo
CC1=CC2=C(C=C1)OC(=CC2=O)C(=O)O
204.181
204.18
≥98.0% (HPLC)
258°C
5006-44-0
MFCD00239435
RKIDLZFIIVDZNX-UHFFFAOYSA-N
6-methyl-4-oxochromene-2-carboxylic acid
688704
Crystal-Powder at 20°C
6-Methylchromone-2-carboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No