Please login to your online account to display your discounted pricing

6-Methyl-5-nitroquinoline, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 6-Methyl-5-nitroquinoline
CAS 23141-61-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H8N2O2
Formula Weight 188.19
MDL Number MFCD00024021
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

BTB10313DA

  SDS
fisher scientific company
BTB10313DA
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 188.19
Quantity 1g
Assay 97%
CAS Max % 100.0
Molecular Formula C10H8N2O2
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00024021
CAS Min % 96.5
Chemical Name or Material 6-Methyl-5-nitroquinoline
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Causes serious eye irritation.
Assay Percent Range 97%
CAS 23141-61-9
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Causes serious eye irritation.