missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methyl-5-nitroquinoline, 97%

$38.70 - $133.85

Chemical Identifiers

CAS 23141-61-9
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD00024021
InChI Key CENBTULRDDPOGR-UHFFFAOYSA-N
Synonym 5-nitro-6-methylquinoline, quinoline, 6-methyl-5-nitro, quinoline,6-methyl-5-nitro, acmc-1cuis, maybridge1_002137, 5-nitro-6-met hylquinoline, 5-nitro-6-methyl quinoline, 6-methyl-5-nitroquinoline
PubChem CID 90009
IUPAC Name 6-methyl-5-nitroquinoline
SMILES CC1=C(C2=C(C=C1)N=CC=C2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0080703 View Documents Alfa Aesar™
L0080703
1g Each for $38.70
 
AAL0080706 View Documents Alfa Aesar™
L0080706
5g Each for $133.85
 
Specifications

Chemical Identifiers

23141-61-9
188.186
CENBTULRDDPOGR-UHFFFAOYSA-N
90009
CC1=C(C2=C(C=C1)N=CC=C2)[N+](=O)[O-]
C10H8N2O2
MFCD00024021
5-nitro-6-methylquinoline, quinoline, 6-methyl-5-nitro, quinoline,6-methyl-5-nitro, acmc-1cuis, maybridge1_002137, 5-nitro-6-met hylquinoline, 5-nitro-6-methyl quinoline, 6-methyl-5-nitroquinoline
6-methyl-5-nitroquinoline

Specifications

115°C to 118°C
23141-61-9
MFCD00024021
5-nitro-6-methylquinoline, quinoline, 6-methyl-5-nitro, quinoline,6-methyl-5-nitro, acmc-1cuis, maybridge1_002137, 5-nitro-6-met hylquinoline, 5-nitro-6-methyl quinoline, 6-methyl-5-nitroquinoline
CC1=C(C2=C(C=C1)N=CC=C2)[N+](=O)[O-]
188.186
188.19
6-Methyl-5-nitroquinoline
1g
C10H8N2O2
154688
CENBTULRDDPOGR-UHFFFAOYSA-N
6-methyl-5-nitroquinoline
90009
97%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger