missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methyl-4-trifluoromethyl-2(1H)-pyridone, 97%

Manufacturer:  Alfa Aesar™ 4346703

 View more versions of this product

Catalog No. AA4346703



Specifications

Specifications

6-Methyl-4-trifluoromethyl-2 (1H)-pyridone
1g
C7H6F3NO
2 1h-pyridinone,6-methyl-4-trifluoromethyl
CC1=CC(=CC(=O)N1)C(F)(F)F
177.126
177.13
97%
129°C to 130°C
22123-19-9
MFCD00042479
CDRMKCNOGIBXOG-UHFFFAOYSA-N
6-methyl-4-(trifluoromethyl)-1H-pyridin-2-one
2782992
Powder

Chemical Identifiers

22123-19-9
177.126
CDRMKCNOGIBXOG-UHFFFAOYSA-N
2782992
CC1=CC(=CC(=O)N1)C(F)(F)F
C7H6F3NO
MFCD00042479
2 1h-pyridinone,6-methyl-4-trifluoromethyl
6-methyl-4-(trifluoromethyl)-1H-pyridin-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents