missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methyl-4-phenyl-2-chromanone, TCI America™

$71.81 - $210.84

Chemical Identifiers

CAS 40546-94-9
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD03695019
InChI Key SUHIZPDCJOQZLN-UHFFFAOYSA-N
Synonym 3,4-Dihydro-6-methyl-4-phenylcoumarin
PubChem CID 3791643
IUPAC Name 6-methyl-4-phenyl-3,4-dihydrochromen-2-one
SMILES CC1=CC2=C(C=C1)OC(=O)CC2C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M209325G View Documents TCI America
M209325G
25g Each for $210.84
 
M20935G View Documents TCI America
M20935G
5g Each for $71.81
 
Specifications

Chemical Identifiers

40546-94-9
238.286
SUHIZPDCJOQZLN-UHFFFAOYSA-N
3791643
CC1=CC2=C(C=C1)OC(=O)CC2C3=CC=CC=C3
C16H14O2
MFCD03695019
3,4-Dihydro-6-methyl-4-phenylcoumarin
6-methyl-4-phenyl-3,4-dihydrochromen-2-one

Specifications

230°C
82°C
40546-94-9
MFCD03695019
SUHIZPDCJOQZLN-UHFFFAOYSA-N
6-methyl-4-phenyl-3,4-dihydrochromen-2-one
3791643
Crystal-Powder at 20°C
6-Methyl-4-phenyl-2-chromanone
White
25g
C16H14O2
3,4-Dihydro-6-methyl-4-phenylcoumarin
CC1=CC2=C(C=C1)OC(=O)CC2C3=CC=CC=C3
238.286
238.29
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No