Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methyl-3-pyridyl trifluoromethanesulfonate, 96%

$111.45 - $378.66

Chemical Identifiers

Chemical Name or Material 6-Methyl-3-pyridyl trifluoromethanesulfonate
CAS 111770-91-3
Molecular Formula C7H6F3NO3S
Formula Weight 241.19
MDL Number MFCD06798124
Synonym 2-Picolin-5-yl trifluoromethanesulfonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2044103

  SDS
alfa aesar™
L2044103
1g Each for $111.45

AAL2044106

  SDS
alfa aesar™
L2044106
5g Each for $378.66
Description & Specifications

Specifications

Formula Weight 241.19
Chemical Name or Material 6-Methyl-3-pyridyl trifluoromethanesulfonate
Molecular Formula C7H6F3NO3S
MDL Number MFCD06798124
Synonym 2-Picolin-5-yl trifluoromethanesulfonate
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 96%
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
CAS 111770-91-3
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.