missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methyl-3-pyridinecarboxaldehyde, TCI America™

$87.08 - $302.50

Chemical Identifiers

CAS 53014-84-9
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD08272279
InChI Key IMWMEIWYPWVABQ-UHFFFAOYSA-N
Synonym 6-methylnicotinaldehyde, 2-methyl-5-formylpyridine, 6-methyl-pyridine-3-carbaldehyde, 3-formyl-6-methyl-pyridine, 5-formyl-2-methylpyridine, 5-formyl-2-picoline, 6-methylpyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-methyl, 3-formyl-6-methylpyridine, 6-methylpyridyl-3-carboxaldehyde
PubChem CID 10192566
IUPAC Name 6-methylpyridine-3-carbaldehyde
SMILES CC1=NC=C(C=C1)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M2654200MG View Documents TCI America
M2654200MG
200mg Each for $87.08
 
M26541G View Documents TCI America
M26541G
1g Each for $302.50
 
Specifications

Chemical Identifiers

53014-84-9
121.139
IMWMEIWYPWVABQ-UHFFFAOYSA-N
10192566
CC1=NC=C(C=C1)C=O
C7H7NO
MFCD08272279
6-methylnicotinaldehyde, 2-methyl-5-formylpyridine, 6-methyl-pyridine-3-carbaldehyde, 3-formyl-6-methyl-pyridine, 5-formyl-2-methylpyridine, 5-formyl-2-picoline, 6-methylpyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-methyl, 3-formyl-6-methylpyridine, 6-methylpyridyl-3-carboxaldehyde
6-methylpyridine-3-carbaldehyde

Specifications

95°C
200mg
C7H7NO
6-methylnicotinaldehyde, 2-methyl-5-formylpyridine, 6-methyl-pyridine-3-carbaldehyde, 3-formyl-6-methyl-pyridine, 5-formyl-2-methylpyridine, 5-formyl-2-picoline, 6-methylpyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-methyl, 3-formyl-6-methylpyridine, 6-methylpyridyl-3-carboxaldehyde
CC1=NC=C(C=C1)C=O
121.139
121.14
≥98.0% (GC)
Undesignated
53014-84-9
MFCD08272279
IMWMEIWYPWVABQ-UHFFFAOYSA-N
6-methylpyridine-3-carbaldehyde
10192566
Clear Liquid at 20°C
6-Methyl-3-pyridinecarboxaldehyde
Safety and Handling

Safety and Handling

TSCA : No