Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methyl-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-propionitrile, 97%

$55.11 - $55.11

Chemical Identifiers

Chemical Name or Material 6-Methyl-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-propionitrile
CAS 351003-19-5
Molecular Formula C12H12N2O2
Formula Weight 216.24
MDL Number MFCD03427105
Synonym 3-[6-Methyl-2H-1,4-benzodioxazin-3(4H)-one-4-yl]propionitrile; 2,3-Dihydro-6-methyl-3-oxo-4H-1,4-benzoxazine-4-propionitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2719803

  SDS
alfa aesar™
H2719803
1g Each for $55.11
Description & Specifications

Specifications

Formula Weight 216.24
Melting Point 128° to 130°C
Chemical Name or Material 6-Methyl-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-propionitrile
Assay 97%
Molecular Formula C12H12N2O2
MDL Number MFCD03427105
CAS 351003-19-5
Synonym 3-[6-Methyl-2H-1,4-benzodioxazin-3(4H)-one-4-yl]propionitrile; 2,3-Dihydro-6-methyl-3-oxo-4H-1,4-benzoxazine-4-propionitrile
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.