missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Methyl-2-pyridinemethanol 98.0+%, TCI America™
Supplier: TCI America M128225G
Specifications
6-Methyl-2-pyridinemethanol | |
White-Yellow | |
MFCD00023520 | |
6-methyl-2-pyridinemethanol, 6-methylpyridin-2-yl methanol, 2-pyridinemethanol, 6-methyl, 6-methyl-2-pyridylmethanol, 6-methylpyridine-2-methanol, 2-hydroxymethyl-6-methylpyridine, 6-methyl-2-pyridyl methan-1-ol, pubchem20959, acmc-2099dw, 6-methyl-2-pyridylcarbinol | |
CC1=CC=CC(CO)=N1 | |
123.16 | |
123.16 | |
Crystalline Lumps |
1122-71-0 | |
C7H9NO | |
25 g | |
JLVBSBMJQUMAMW-UHFFFAOYSA-N | |
(6-methylpyridin-2-yl)methanol | |
70736 | |
≥98.0% (GC) |
Chemical Identifiers
1122-71-0 | |
123.16 | |
JLVBSBMJQUMAMW-UHFFFAOYSA-N | |
70736 | |
CC1=CC=CC(CO)=N1 |
C7H9NO | |
MFCD00023520 | |
6-methyl-2-pyridinemethanol, 6-methylpyridin-2-yl methanol, 2-pyridinemethanol, 6-methyl, 6-methyl-2-pyridylmethanol, 6-methylpyridine-2-methanol, 2-hydroxymethyl-6-methylpyridine, 6-methyl-2-pyridyl methan-1-ol, pubchem20959, acmc-2099dw, 6-methyl-2-pyridylcarbinol | |
(6-methylpyridin-2-yl)methanol |
Safety and Handling
TSCA : No