Learn More
6-Methyl-2-pyridinemethanol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2679406
Description
Use as primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUse as primary and secondary intermediate.
Solubility
Low soluble in water, high soluble in ethanol, acetic acid. Soluble in benzene, toluene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, strong reducing agents. Store under an inert atmosphere.
Specifications
6-Methyl-2-pyridinemethanol | |
32°C to 34°C | |
109°C (228°F) | |
MFCD00023520 | |
6-methyl-2-pyridinemethanol, 6-methylpyridin-2-yl methanol, 2-pyridinemethanol, 6-methyl, 6-methyl-2-pyridylmethanol, 6-methylpyridine-2-methanol, 2-hydroxymethyl-6-methylpyridine, 6-methyl-2-pyridyl methan-1-ol, pubchem20959, acmc-2099dw, 6-methyl-2-pyridylcarbinol | |
JLVBSBMJQUMAMW-UHFFFAOYSA-N | |
(6-methylpyridin-2-yl)methanol | |
70736 | |
98% |
1122-71-0 | |
105°C to 108°C (12 mmHg) | |
C7H9NO | |
5 g | |
Low soluble in water,high soluble in ethanol,acetic acid. Soluble in benzene,toluene. | |
CC1=CC=CC(CO)=N1 | |
123.16 | |
123.15 |
Chemical Identifiers
1122-71-0 | |
123.16 | |
JLVBSBMJQUMAMW-UHFFFAOYSA-N | |
70736 | |
CC1=CC=CC(CO)=N1 |
C7H9NO | |
MFCD00023520 | |
6-methyl-2-pyridinemethanol, 6-methylpyridin-2-yl methanol, 2-pyridinemethanol, 6-methyl, 6-methyl-2-pyridylmethanol, 6-methylpyridine-2-methanol, 2-hydroxymethyl-6-methylpyridine, 6-methyl-2-pyridyl methan-1-ol, pubchem20959, acmc-2099dw, 6-methyl-2-pyridylcarbinol | |
(6-methylpyridin-2-yl)methanol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-358-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only