Please login to your online account to display your discounted pricing

6-Methyl-2-pyridinemethanol, +95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 6-Methyl-2-pyridinemethanol
CAS 1122-71-0
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 95+%
Molecular Formula C7H9NO
Formula Weight 123.15
MDL Number MFCD00023520
Synonym (6-Methylpyridin-2-yl)methanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC393420010

  SDS
thermo fisher scientific
AC393420010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95.0%
Physical Form Liquid or Low Melting Solid
Formula Weight 123.15
Quantity 1g
Color Clear light brown
CAS Min % 95.0
CAS Max % 100.0
Synonym (6-Methylpyridin-2-yl)methanol
Molecular Formula C7H9NO
Assay Percent Range 95+%
MDL Number MFCD00023520
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if swallowed.
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 1122-71-0
Chemical Name or Material 6-Methyl-2-pyridinemethanol
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if swallowed.