missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methyl-2-(p-tolylamino)-4H-3,1-benzoxazin-4-one, TCI America™

$190.97 - $667.66

Chemical Identifiers

CAS 86672-58-4
Molecular Formula C16H14N2O2
Molecular Weight (g/mol) 266.3
MDL Number MFCD03272508
InChI Key GFWNGVKCDGYFKG-UHFFFAOYSA-N
Synonym URB-754, 6-Methyl-2-(p-tolylamino)-4H-benzo[d][1,3]oxazin-4-one
PubChem CID 848487
IUPAC Name 6-methyl-2-(4-methylanilino)-3,1-benzoxazin-4-one
SMILES CC1=CC=C(C=C1)NC2=NC3=C(C=C(C=C3)C)C(=O)O2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M257210MG View Documents TCI America
M257210MG
10mg Each for $190.97
 
M257250MG View Documents TCI America
M257250MG
50mg Each for $667.66
 
Specifications

Chemical Identifiers

86672-58-4
266.3
GFWNGVKCDGYFKG-UHFFFAOYSA-N
848487
CC1=CC=C(C=C1)NC2=NC3=C(C=C(C=C3)C)C(=O)O2
C16H14N2O2
MFCD03272508
URB-754, 6-Methyl-2-(p-tolylamino)-4H-benzo[d][1,3]oxazin-4-one
6-methyl-2-(4-methylanilino)-3,1-benzoxazin-4-one

Specifications

Yellow
10mg
C16H14N2O2
URB-754, 6-Methyl-2-(p-tolylamino)-4H-benzo[d][1,3]oxazin-4-one
CC1=CC=C(C=C1)NC2=NC3=C(C=C(C=C3)C)C(=O)O2
266.3
266.30
≥95.0% (HPLC)
215°C
86672-58-4
MFCD03272508
GFWNGVKCDGYFKG-UHFFFAOYSA-N
6-methyl-2-(4-methylanilino)-3,1-benzoxazin-4-one
848487
Crystal-Powder at 20°C
6-Methyl-2-(p-tolylamino)-4H-3,1-benzoxazin-4-one
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer (-20 °C)