Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methyl-2-heptyne, 98%

$36.32 - $36.32

Chemical Identifiers

Chemical Name or Material 6-Methyl-2-heptyne
CAS 51065-64-6
Molecular Formula (CH3)2CHCH2CH2C≡CCH3
Formula Weight 110.2
MDL Number MFCD00041619
UN Number UN3295
Synonym Methyl isopentyl acetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2088103

  SDS
alfa aesar™
B2088103
1g Each for $36.32

AAB2088106

  SDS
alfa aesar™
B2088106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2088114

  SDS
alfa aesar™
B2088114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 110.2
Chemical Name or Material 6-Methyl-2-heptyne
Assay 98%
Molecular Formula (CH3)2CHCH2CH2C≡CCH3
MDL Number MFCD00041619
CAS 51065-64-6
UN Number UN3295
Synonym Methyl isopentyl acetylene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.