missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methyl-2-heptanone, TCI America™

$149.72 - $433.89

Chemical Identifiers

CAS 928-68-7
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00026527
InChI Key DPLGXGDPPMLJHN-UHFFFAOYSA-N
Synonym Methyl 4-Methylpentyl Ketone
PubChem CID 13572
IUPAC Name 6-methylheptan-2-one
SMILES CC(C)CCCC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M07005ML View Documents TCI America
M07005ML
5mL Each for $149.72
 
M070025ML View Documents TCI America
M070025ML
25mL Each for $433.89
 
Specifications

Chemical Identifiers

928-68-7
128.215
DPLGXGDPPMLJHN-UHFFFAOYSA-N
13572
CC(C)CCCC(=O)C
C8H16O
MFCD00026527
Methyl 4-Methylpentyl Ketone
6-methylheptan-2-one

Specifications

171°C
5mL
C8H16O
1224
DPLGXGDPPMLJHN-UHFFFAOYSA-N
6-methylheptan-2-one
13572
Clear Liquid at 20°C
6-Methyl-2-heptanone
Yellow
928-68-7
MFCD00026527
Methyl 4-Methylpentyl Ketone
CC(C)CCCC(=O)C
128.215
128.22
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0542

TSCA : Yes