Please login to your online account to display your discounted pricing

6-Methyl-2,4-heptanedione, 98%, ACROS Organics™

$371.67 - $371.67

Chemical Identifiers

Chemical Name or Material 6-Methyl-2, 4-heptanedione
Name Note 98%
CAS 3002-23-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8 H14 O2
Linear Formula CH3COCH2COCH2CH(CH3)2
Formula Weight 142.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC414400250

  SDS
thermo fisher scientific
414400250
Glass bottle 25g Each for $371.67
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 142.2
Physical Form Liquid
Quantity 25g
Color Clear colorless to pale yellow
TSCA TSCA
EINECS Number 221-093-6
Density 0.9200g/mL
CAS Max % 100.0
Packaging Glass bottle
CAS Min % 97.5
Refractive Index 1.4550 to 1.4577 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Warning
Chemical Name or Material 6-Methyl-2, 4-heptanedione
Name Note 98%
Assay Percent Range 98%
Linear Formula CH3COCH2COCH2CH(CH3)2
MDL Number MFCD00008939
CAS 3002-23-1
Molecular Formula C8 H14 O2
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.