Please login to your online account to display your discounted pricing

6-methyl-2,3-dihydropyridazin-3-one hydrate, Maybridge


Quantity 50g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  BTB06840EE

 View more versions of this product

Catalog No. BTB06840EE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 128.14
Quantity 50g
Assay 90%
Chemical Name or Material 6-Methyl-2, 3-dihydropyridazin-3-one hydrate
Health Hazard 1 Warning
Packaging Amber glass bottle
Assay Percent Range 90%
Molecular Formula C5H6N2O·H2O
CAS 7143-82-0

Specifications

Chemical Name or Material 6-Methyl-2, 3-dihydropyridazin-3-one hydrate
Assay 90%
Assay Percent Range 90%
Molecular Formula C5H6N2O·H2O
Formula Weight 128.14
Accessories

Warning