missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methyl-1-indanone, 98%

$66.00 - $212.81

Chemical Identifiers

CAS 24623-20-9
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.189
MDL Number MFCD00275729
InChI Key DBOXRDYLMJMQBB-UHFFFAOYSA-N
Synonym 6-methyl-1-indanone, 6-methyl-2,3-dihydro-1h-inden-1-one, 6-methylindan-1-one, 6-methyl-indan-1-one, 1h-inden-1-one, 2,3-dihydro-6-methyl, 2,3-dihydro-6-methylinden-1-one, pubchem8851, acmc-209gdm, 6-methyl-indane-1-one, 6-methyl-1-indanone
PubChem CID 2795093
IUPAC Name 6-methyl-2,3-dihydroinden-1-one
SMILES CC1=CC2=C(CCC2=O)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1755903 View Documents Alfa Aesar™
L1755903
1g Each for $66.00
 
AAL1755906 View Documents Alfa Aesar™
L1755906
5g Each for $212.81
 
Specifications

Chemical Identifiers

24623-20-9
146.189
DBOXRDYLMJMQBB-UHFFFAOYSA-N
2795093
CC1=CC2=C(CCC2=O)C=C1
C10H10O
MFCD00275729
6-methyl-1-indanone, 6-methyl-2,3-dihydro-1h-inden-1-one, 6-methylindan-1-one, 6-methyl-indan-1-one, 1h-inden-1-one, 2,3-dihydro-6-methyl, 2,3-dihydro-6-methylinden-1-one, pubchem8851, acmc-209gdm, 6-methyl-indane-1-one, 6-methyl-1-indanone
6-methyl-2,3-dihydroinden-1-one

Specifications

124°C to 126°C (11mmHg)
1g
C10H10O
1448005
DBOXRDYLMJMQBB-UHFFFAOYSA-N
6-methyl-2,3-dihydroinden-1-one
2795093
98%
57°C to 59°C
24623-20-9
MFCD00275729
6-methyl-1-indanone, 6-methyl-2,3-dihydro-1h-inden-1-one, 6-methylindan-1-one, 6-methyl-indan-1-one, 1h-inden-1-one, 2,3-dihydro-6-methyl, 2,3-dihydro-6-methylinden-1-one, pubchem8851, acmc-209gdm, 6-methyl-indane-1-one, 6-methyl-1-indanone
CC1=CC2=C(CCC2=O)C=C1
146.189
146.19
6-Methyl-1-indanone